Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP17 Antibody (OTI2G6) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274307
Description
SP17 Monoclonal specifically detects SP17 in Human samples. It is validated for Western Blot.Specifications
SP17 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
Cancer/testis antigen 22, CT22SP17-1, Sp17-1, SP17sperm surface protein Sp17, sperm autoantigenic protein 17Sperm protein 17 | |
Mouse | |
17.2 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI2G6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SPA17 | |
Full length human recombinant protein of human SPA17 (NP_059121) produced in E.coli. | |
Affinity Purified | |
Cancer | |
53340 | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction