Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP17 Antibody (OTI2G6), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274307H
Description
SP17 Monoclonal specifically detects SP17 in Human samples. It is validated for Western Blot.Specifications
SP17 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SPA17 | |
Full length recombinant protein of human SP17 (NP_059121) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2G6 | |
Western Blot | |
Cancer/testis antigen 22, CT22SP17-1, Sp17-1, SP17sperm surface protein Sp17, sperm autoantigenic protein 17Sperm protein 17 | |
Mouse | |
Affinity Purified | |
Cancer | |
53340.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction