Learn More
Invitrogen™ SP3 Polyclonal Antibody

Description
PA1-30616 detects Sp3 from mouse, rat, human samples.
Specifications
Specifications
| Antigen | SP3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% gelatin and 0.1% sodium azide |
| Gene | SP3 |
| Gene Accession No. | O70494, Q02447 |
| Gene Alias | D130027J01Rik; DKFZp686O1631; GC-binding transcription factor Sp3; SP3; Sp3 transcription factor; specificity protein 3; SPR1; SPR-1; SPR2; SPR-2; trans-acting transcription factor 3; transcription factor Sp3 |
| Gene Symbols | SP3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.