Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SP3/SP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599719
Description
Antibody detects endogenous levels of total Sp3/4.
This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13. [provided by RefSeq, Feb 2010].
Specifications
SP3/SP4 | |
Polyclonal | |
Unconjugated | |
SP4 | |
5730497N03Rik; D130027J01Rik; DKFZp686O1631; GC-binding transcription factor Sp3; HF1B; HF-1b; HF1-b; HF-1b zinc finger protein; SP3; Sp3 transcription factor; SP4; Sp4 transcription factor; specificity protein 3; SPR1; SPR-1; SPR2; SPR-2; trans-acting transcription factor 3; trans-acting transcription factor 4; transcription factor Sp3; Transcription factor Sp4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
6670, 6671 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q02446, Q02447 | |
SP3, SP4 | |
A synthesized peptide derived from human SP3(Accession Q02447), corresponding to amino acid residues R639-R689. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction