Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ SP4 (Human) Recombinant Protein
Shop All Abnova Corporation Products
Click to view available options
Quantity:
2 μg
Description
Human SP4 full-length ORF (ABZ92432.1, 1 a.a. - 784 a.a.) recombinant protein with GST tag at N-terminal.
Specifications
Specifications
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Gene ID (Entrez) | 6671 |
Name | Sp4 transcription factor |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue |
Quantity | 2 μg |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Gene Alias | HF1B, MGC130008, MGC130009, SPR-1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction