Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595764
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595764 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595764 Supplier Invitrogen™ Supplier No. PA595764
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGQVGQPAAT ADSGTSNGNQ LVSTPTNTTT SASTMPESPS SSTTCTTTAS TSLTSSDTLV SSADTGQYAS TSASSSERTI EESQTPAATE SEAQSSSQLQ PNGMQNAQDQ SNSLQQVQIV GQPILQQIQI QQPQQQIIQA IPPQSFQLQS GQTIQTIQQQ P; Positive Samples: Mouse thymus, Mouse spleen, Mouse pancreas, Rat thymus; Cellular Location: Nucleus.

This gene belongs to a family of Sp1 related genes that encode transcription factors that regulate transcription by binding to consensus GC- and GT-box regulatory elements in target genes. This protein contains a zinc finger DNA-binding domain and several transactivation domains, and has been reported to function as a bifunctional transcription factor that either stimulates or represses the transcription of numerous genes. Transcript variants encoding different isoforms have been described for this gene, and one has been reported to initiate translation from a non-AUG (AUA) start codon. Additional isoforms, resulting from the use of alternate downstream translation initiation sites, have also been noted. A related pseudogene has been identified on chromosome 13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Sp4
Gene Accession No. Q02446, Q62445
Gene Alias 5730497N03Rik; HF1B; HF-1b; HF1-b; HF-1b zinc finger protein; LOW QUALITY PROTEIN: transcription factor Sp4; MGC130008; MGC130009; SP4; Sp4 transcription factor; SPR-1; trans-acting transcription factor 4; transcription factor Sp4
Gene Symbols Sp4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 280-440 of human SP4 (NP_0031032).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20688, 25162, 6671
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.