Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543649
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543649 Supplier Invitrogen™ Supplier No. PA543649
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DHLAKHMKTH EGAKEEAAGA ASGEGKAGGA VEPPGGKGKR EAEGSVAPSN Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.

SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Sp6
Gene Accession No. Q3SY56
Gene Alias 1110025J03Rik; AA591031; AI592962; EPFN; EPIPROFIN; Klf14; krueppel-like factor 14; Kruppel-like factor 14; LOW QUALITY PROTEIN: transcription factor Sp6; MGC119662; MGC119663; MGC119664; MGC119665; SP6; Sp6 transcription factor; specificity protein 6; trans-acting transcription factor 6; transcription factor Sp6
Gene Symbols Sp6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SP6 (aa 327-376).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80320
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.