Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115955
Description
Antibody detects endogenous levels of total SP6.
SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).
Specifications
| SP6 | |
| Polyclonal | |
| Unconjugated | |
| Sp6 | |
| 1110025J03Rik; AA591031; AI592962; EPFN; EPIPROFIN; Klf14; krueppel-like factor 14; Kruppel-like factor 14; LOW QUALITY PROTEIN: transcription factor Sp6; MGC119662; MGC119663; MGC119664; MGC119665; SP6; Sp6 transcription factor; specificity protein 6; trans-acting transcription factor 6; transcription factor Sp6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80320, 83395 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q3SY56, Q9ESX2 | |
| Sp6 | |
| A synthesized peptide derived from human SP6(Accession Q3SY56), corresponding to amino acid residues P91-S141. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction