Learn More
Thermo Scientific™ SP6 RNA Polymerase (20 U/μL) PROMO
SP6 RNA Polymerase is a DNA-dependent RNA polymerase with strict specificity for its double-stranded promoter.
Supplier:  Thermo Scientific™ EP0131
Catalog No. FEREP0131PM
Description
Description
It catalyzes the 5' => 3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter.
  • Incorporates modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)
Recommended for:
Synthesis of unlabeled and labeled RNA that can be used: for hybridization (1), in vitro RNA translation (2); as aRNA (3), siRNA (4), substrate in RNase protection assays (5), template for genomic DNA sequencing.
Note:
Consensus promoter sequences: AATTAACCCTCACTAAA G GGAGA - T3 TAATACGACTCACTATA G GGAGA - T7 ATTTAGGTGACACTATA G AAGNG - SP6 The position in bold (+1) indicates the first nucleotide incorporated into RNA during transcription. Only bases at this position through +3 are critical for transcription, and they must be a G and a purine base, respectively (9).
Specifications
Specifications
SP6 RNA Polymerase
2000 Units
1X
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only. Not for use in diagnostic procedures.