Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116893
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116893 Supplier Invitrogen™ Supplier No. PA5116893
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Rat testis, Rat brain Immunogen sequence: KRFTRSDELE RHVRTHTREK KFTCLLCSKR FTRSDHLSKH QRTH.

SP7 is a C2H2-type zinc finger transcription factor of the SP gene family and a putative master regulator of bone cell differentiation (Gao et al., 2004 [PubMed 15474293]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene SP7
Gene Accession No. Q8TDD2, Q8VI67
Gene Alias 6430578P22Rik; C22; MGC126598; OI11; OI12; osterix; OSX; Sp7; Sp7 transcription factor; Sp7 transcription factor 7; trans-acting transcription factor 7; transcription factor Sp7; Zinc finger protein osterix
Gene Symbols SP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 333-376 of human SP7 (NP_0011669381).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121340, 170574, 300260
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.