Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524289
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524289 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524289 Supplier Invitrogen™ Supplier No. PA524289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with chicken, Xenopus and zebrafish based on sequence homology.

Transcription factor which plays a key role in limb development. Positively regulates FGF8 expression in the apical ectodermal ridge (AER) and contributes to limb outgrowth in embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SP8
Gene Accession No. Q8BMJ8, Q8IXZ3
Gene Alias BTD; D930049B17Rik; mBtd; Sp8; Sp8 transcription factor; specificity protein 8; trans-acting transcription factor 8; transcription factor Sp8
Gene Symbols SP8
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 386-415 amino acids from the C-terminal region of human SP8.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221833, 320145
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.