Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570643 Supplier Invitrogen™ Supplier No. PA570643
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.

Transcription factor which plays a key role in limb development. Positively regulates FGF8 expression in the apical ectodermal ridge (AER) and contributes to limb outgrowth in embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SP9
Gene Accession No. P0CG40
Gene Alias buttonhead-like zinc finger transcription factor; buttonhead-like zinc finger transcription factor Sp9; RGD1559457; Sp9; sp9 protein; Sp9 transcription factor; Sp9 transcription factor homolog; trans-acting transcription factor 9; transcription factor Sp9; zgc:85876; zinc finger protein 990; zinc finger transcription factor; ZNF990
Gene Symbols SP9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SP9 (aa 414-463).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131390
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.