Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564038
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564038 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564038 Supplier Invitrogen™ Supplier No. PA564038
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKHIKTHNGG GGGKKGSDSD TDASNLETPR SESPDLILHD SGVSA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Transcription factor which plays a key role in limb development. Positively regulates FGF8 expression in the apical ectodermal ridge (AER) and contributes to limb outgrowth in embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP9
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SP9
Gene Accession No. P0CG40
Gene Alias buttonhead-like zinc finger transcription factor; buttonhead-like zinc finger transcription factor Sp9; RGD1559457; Sp9; sp9 protein; Sp9 transcription factor; Sp9 transcription factor homolog; trans-acting transcription factor 9; transcription factor Sp9; zgc:85876; zinc finger protein 990; zinc finger transcription factor; ZNF990
Gene Symbols SP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SP9. Recombinant protein control fragment (Product #RP-103457).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131390
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.