Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPA17 Monoclonal Antibody (1C4C7)
GREENER_CHOICE

Catalog No. FERMA548752
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548752 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548752 Supplier Invitrogen™ Supplier No. MA548752
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPA17 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPA17
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1C4C7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SPA17
Gene Accession No. Q15506
Gene Alias band 34; cancer/testis antigen 22; CT22; SP17; SP17 protein; SP17-1; SPA17; Sperm autoantigenic protein 17; sperm protein 17; Sperm surface protein Sp17; Unknown (protein for MGC:127855); zgc:153721
Gene Symbols SPA17
Host Species Mouse
Immunogen Purified recombinant fragment of human SP17 (AA: 1-151) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53340
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.