Learn More
SPA17 Mouse anti-Human, Clone: OTI4A2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804222

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a protein present at the cell surface. The N-terminus has sequence similarity to human cAMP-dependent protein kinase A (PKA) type II alpha regulatory subunit (RIIa) while the C-terminus has an IQ calmodulin-binding motif. The central portion of the protein has carbohydrate binding motifs and likely functions in cell-cell adhesion. The protein was initially characterized by its involvement in the binding of sperm to the zona pellucida of the oocyte. Recent studies indicate that it is also involved in additional cell-cell adhesion functions such as immune cell migration and metastasis. A retrotransposed pseudogene is present on chromosome 10q22.Specifications
SPA17 | |
Monoclonal | |
Unconjugated | |
SPA17 | |
CT22, SP17, SP17-1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
53340 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15506 | |
SPA17 | |
Full length human recombit protein of human SPA17 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.