Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAC7 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989195
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9195 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9195 Supplier Bioss Supplier No. BS9608R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPACA7 is a protein coding gene involved in fertilization. Seems not to play a direct role in sperm-egg binding or gamete fusion.

Specifications

Antigen SPAC7
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPACA7
Gene Accession No. Q96KW9
Gene Alias C13orf28; protein SPACA7; SPACA7; sperm acrosome associated 7; Sperm acrosome-associated protein 7
Gene Symbols SPACA7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPAC7/C13ORF28.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 122258
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.