Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPACA1 Polyclonal Antibody

SPACA1 Polyclonal Antibody
Supplier: Thermo Scientific PA555400
Description
SPACA1 Polyclonal Antibody for Western Blot, IHC

Specifications
SPACA1 | |
Polyclonal | |
Unconjugated | |
SPACA1 | |
1700124L11Rik; 4930540L03Rik; RGD1560284; SAMP32; SPACA1; Sperm acrosomal membrane-associated protein 32; sperm acrosome associated 1; sperm acrosome membrane-associated protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81833 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBV2 | |
SPACA1 | |
Recombinant protein corresponding to Human SPACA1. Recombinant protein control fragment (Product #RP-94711). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction