Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPACA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502209213
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-9213 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-9213 Supplier Fabgennix Inc Supplier No. SPACA3331AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPACA3 Polyclonal antibody specifically detects SPACA3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen SPACA3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene SPACA3
Gene Accession No. Q8IXA5, Q9D9X8
Gene Alias 1700025M08Rik; acrosomal sperm protein; ALLP17; Cancer/testis antigen 54; CT54; LYC3; lysozyme C; lysozyme-like acrosomal sperm-specific secretory protein ALLP-17; Lysozyme-like protein 3; lysozyme-like sperm-specific secretory protein ALLP17; LYZC; Lyzl3; mSLLP1; SLLP1; Spaca3; sperm acrosome associated 3; sperm acrosome membrane-associated protein 3; Sperm acrosome membrane-associated protein 3, membrane form; Sperm acrosome membrane-associated protein 3, processed form; sperm c-type lysozyme-like protein; sperm lysozyme like protein 1; sperm lysozyme-like protein 1; sperm lysozyme-like protein 1 precusor; sperm lyzozyme-like acrosomal protein 1; Sperm protein reactive with antisperm antibodies; Sperm protein reactive with ASA; SPRASA; UNQ424/PRO862
Host Species Rabbit
Immunogen Synthetic peptides taken within amino acid region 50-100 and 80-130 on human sperm acrosome membrane-associated protein 3 isoform 2.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124912, 287557, 75622
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.