Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPACA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559791
Description
Immunogen sequence: MHCGDDEDCF TGHGVAPGTG PVINKGCLRA TSCGLEEPVS YRGVTYSLTT NCCTGRLCNR APSSQTVGAT TSLALGLGML LPPR Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 61%.

Specifications
SPACA4 | |
Polyclonal | |
Unconjugated | |
SPACA4 | |
SAMP14; SPACA4; sperm acrosomal membrane protein 14; sperm acrosomal membrane-associated protein 14; sperm acrosome associated 4; sperm acrosome membrane-associated protein 4; testicular tissue protein Li 167; UNQ3046/PRO9862 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171169 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDM5 | |
SPACA4 | |
Recombinant protein corresponding to Human SPACA4. Recombinant protein control fragment (Product #RP-99169). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction