Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPACA5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560813 Supplier Invitrogen™ Supplier No. PA560813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AGLNGYKGYG VGDWLCMAHY ESGFDTAFVD HNPDGSSEYG IFQLNSAWWC DNGITPTKNL C Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

SPACA5 (Sperm Acrosome Associated 5) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include lysozyme activity. An important paralog of this gene is SPACA5B. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPACA5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPACA5
Gene Accession No. Q96QH8
Gene Alias Gm718; LYC5; lysozyme-like protein 5; Lyzl5; novel C-type lysozyme/alpha-lactalbumin family domain containing protein; PNPK6288; SLLP2; SLLP-X; Spaca5; SPACA5A; SPACA5B; sperm acrosome associated 5; sperm acrosome-associated protein 5; sperm lysozyme-like protein 2; sperm-specific lysozyme-like protein X; testicular tissue protein Li 113; UNQ6288; UNQ6288/PRO20753
Gene Symbols SPACA5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPACA5. Recombinant protein control fragment (Product #RP-104371).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389852
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.