Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPACA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565674
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565674 Supplier Invitrogen™ Supplier No. PA565674
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAHSEKLPRKV LQHVSEPQAH QESTRGAARP AQAIGTQPRA TKHKCRQLTK ASLKPRGCSK PPWRPPGGKL Highest antigen sequence identity to the following orthologs - mouse 32%, rat 32%.

SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPACA9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPACA9
Gene Accession No. Q96E40
Gene Alias 1700026L06Rik; 80-kDa SOGA fragment; Acrosome and sperm tail protein; C11H9orf9; C20orf117; C9H9orf9; C9orf9; ci009; C-terminal 80 kDa form; KIAA0889; MAST; N-terminal form; Protein SOGA1; RIKEN cDNA 1700026L06 gene; Rsb66; Rsb66 homolog; Rsb-66 protein; SOGA; SOGA family member 1; SOGA1; SPACA9; sperm acrosome associated 9; sperm acrosome-associated protein 9; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1; uncharacterized protein C9orf9 homolog
Gene Symbols SPACA9
Host Species Rabbit
Immunogen Recombinant Human CI009. Recombinant protein control fragment (Product #RP-104927).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11092
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.