Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPACA9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569982
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569982 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569982 Supplier Invitrogen™ Supplier No. PA569982
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 92%; Rat: 100%.

SPACA9 (Sperm Acrosome Associated 9) is a Protein Coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPACA9
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPACA9
Gene Accession No. Q4V8P4
Gene Alias 1700026L06Rik; 80-kDa SOGA fragment; Acrosome and sperm tail protein; C11H9orf9; C20orf117; C9H9orf9; C9orf9; ci009; C-terminal 80 kDa form; KIAA0889; MAST; N-terminal form; Protein SOGA1; RIKEN cDNA 1700026L06 gene; Rsb66; Rsb66 homolog; Rsb-66 protein; SOGA; SOGA family member 1; SOGA1; SPACA9; sperm acrosome associated 9; sperm acrosome-associated protein 9; Suppressor of glucose by autophagy; suppressor of glucose from autophagy; suppressor of glucose, autophagy associated 1; suppressor of glucose, autophagy-associated protein 1; uncharacterized protein C9orf9 homolog
Gene Symbols SPACA9
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human Rsb66 (aa 30-79).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 296610
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.