Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545738 Supplier Invitrogen™ Supplier No. PA545738
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LHVGKEKYSK RPTKKKTPRD YAEWDKFDVE KECLKIDEDY KEKTVIDKSH Sequence homology: Dog: 85%; Human: 100%.

The correlation of anti-sperm antibodies with cases of unexplained infertility implicates a role for these antibodies in blocking fertilization. Improved diagnosis and treatment of immunologic infertility, as well as identification of proteins for targeted contraception, are dependent on the identification and characterization of relevant sperm antigens. The protein expressed by this gene is recognized by anti-sperm agglutinating antibodies from an infertile woman. Furthermore, immunization of female rats with the recombinant human protein reduced fertility. This protein localizes to the plasma membrane of germ cells in the testis and to the post-acrosomal plasma membrane of mature spermatozoa. Recombinant polypeptide binds GTP and exhibits GTPase activity. Thus, this protein may regulate GTP signal transduction pathways involved in spermatogenesis and fertilization. Two transcript variants of this gene encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPAG1
Gene Accession No. Q07617
Gene Alias CILD28; CT140; epididymis secretory protein Li 268; HEL-S-268; HSD-3.8; Infertility-related sperm protein Spag-1; SP75; SPAG1; sperm associated antigen 1; sperm-associated antigen 1; sperm-associated antigen 1; LOW QUALITY PROTEIN: sperm-associated antigen 1; tetratricopeptide repeat-containing protein; TPIS; TPR-containing protein involved in spermatogenesis
Gene Symbols SPAG1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SPAG1 (aa 138-187).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6674
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.