Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524323
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524323 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524323 Supplier Invitrogen™ Supplier No. PA524323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPAG11A Polyclonal Antibody for Western Blot

Sperm binding proteins and their C-terminal peptides of the Sperm Associated Antigen 11 (SPAG11) family are found to play an important role in epididymal innate immunity in addition to their role in sperm maturation. SPAG11 peptides resulting due to cleavage at the furin-like proprotein convertase motif are reported in the epididymal fluid, ejaculate and on sperm and their antimicrobial activity demonstrated. SPAG11A belongs to this family and is also named as EP2, HE2. It has 4 isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG11A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SPAG11A
Gene Accession No. Q6PDA7
Gene Alias 9230111C08Rik; antimicrobial peptide Bin1b; Antimicrobial-like protein Bin-1b; beta defensin EP2e; Bin1b; EDDM2A; EP2; EP2e; EP2Q; epididymal protein 2A; HE2; Human epididymis-specific protein 2; Protein EP2; Spag11; SPAG11A; SPAG11B; Sperm antigen HE2; sperm associated antigen 11; sperm associated antigen 11A; sperm-associated antigen 11; Sperm-associated antigen 11A; Sperm-associated antigen 11B
Gene Symbols SPAG11A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 14-42 amino acids from the N-terminal region of human SPAG11A.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.