Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPAG11A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116883
Description
Positive Samples: Rat testis Immunogen sequence: MRQRLLPSVT SLLLVALLFP GSSQARHVNH SATEALGELR ERAPGQGTNG.
Sperm binding proteins and their C-terminal peptides of the Sperm Associated Antigen 11 (SPAG11) family are found to play an important role in epididymal innate immunity in addition to their role in sperm maturation. SPAG11 peptides resulting due to cleavage at the furin-like proprotein convertase motif are reported in the epididymal fluid, ejaculate and on sperm and their antimicrobial activity demonstrated. SPAG11A belongs to this family and is also named as EP2, HE2. It has 4 isoforms produced by alternative splicing.
Specifications
SPAG11A | |
Polyclonal | |
Unconjugated | |
SPAG11A | |
9230111C08Rik; antimicrobial peptide Bin1b; Antimicrobial-like protein Bin-1b; beta defensin EP2e; Bin1b; EDDM2A; EP2; EP2e; EP2Q; epididymal protein 2A; HE2; Human epididymis-specific protein 2; Protein EP2; Spag11; SPAG11A; SPAG11B; Sperm antigen HE2; sperm associated antigen 11; sperm associated antigen 11A; sperm-associated antigen 11; Sperm-associated antigen 11A; Sperm-associated antigen 11B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
246305, 78128 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8VBV2 | |
SPAG11A | |
A synthetic peptide corresponding to a sequence within amino acids 1-50 of human SPAG11A (NP_4781082). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction