Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582847 Supplier Invitrogen™ Supplier No. PA582847
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVPLGIRNTI CRMQQGICRL FFCHSGEKKR DICSDPWNRC CVSNTDEEGK EKPEMDGRSG I Highest antigen sequence indentity to the following orthologs: Rat - 54%, Mouse - 54%.

SPAG11B is one of several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides.This gene encodes several androgen-dependent, epididymis-specific secretory proteins. The specific functions of these proteins have not been determined, but they are thought to be involved in sperm maturation. Some of the isoforms contain regions of similarity to beta-defensins, a family of antimicrobial peptides. The gene is located on chromosome 8p23 near the defensin gene cluster. Alternative splicing of this gene results in seven transcript variants encoding different isoforms. Two different N-terminal and five different C-terminal protein sequences are encoded by the splice variants. Two additional variants have been described, but their full length sequences have not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG11B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPAG11B
Gene Accession No. Q08648
Gene Alias EDDM2B; EP2; EP2C; EP2D; epididymal protein 2B; HE2; HE2C; Human epididymis-specific protein 2; MGC61846; Protein EP2; SPAG11; SPAG11A; SPAG11B; Sperm antigen HE2; sperm associated antigen 11B; Sperm-associated antigen 11A; Sperm-associated antigen 11B
Gene Symbols SPAG11B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPAG11B. Recombinant protein control fragment (Product #RP-108317).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.