Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142541
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142541 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142541 Supplier Invitrogen™ Supplier No. PA5142541
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognize isoform 1 and 2 (NP_078808.3; NP_001020607.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG16
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene SPAG16
Gene Accession No. Q8K450, Q8N0X2
Gene Alias 4921511D23Rik; 4930524F24Rik; 4930585K05Rik; AV261009; PF20; Pf20 protein homolog; RGD1565062; SPAG16; sperm associated antigen 16; sperm-associated antigen 16 protein; sperm-associated WD repeat protein; spg16; WD repeat domain 29; WDR29
Gene Symbols SPAG16
Host Species Goat
Immunogen Peptide with sequence C-KGVTELRTVGNVPD.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66722, 79582
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.