Learn More
SPAG4 Mouse anti-Human, Clone: OTI3E9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809875

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.Specifications
SPAG4 | |
Monoclonal | |
Unconjugated | |
SPAG4 | |
CT127, SUN4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6676 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3E9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NPE6 | |
SPAG4 | |
Human recombit protein fragment corresponding to amino acids 191-279 of human SPAG4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.