Learn More
Invitrogen™ SPAG4L Polyclonal Antibody

Description
The antibody detects endogenous levels of total SUN5 protein.
Specifications
Specifications
| Antigen | SPAG4L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 2.6 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | SUN5 |
| Gene Accession No. | Q8TC36 |
| Gene Alias | 1700021O15Rik; dJ726C3.1; Sad1 and UNC84 domain containing 5; sad1 and UNC84 domain-containing protein 5; SPAG4L; SPAG4L-2; Sperm-associated antigen 4-like protein; SRG4; SUN domain-containing protein 5; SUN5; testis and spermatogenesis related gene 4; testis and spermatogenesis-related gene 4 protein; TSARG4 |
| Gene Symbols | SUN5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.