Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561761
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561761 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561761 Supplier Invitrogen™ Supplier No. PA561761
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPRSSRSPGD PGALLEDVAH NPRPRRIAQR GRNTSRMAED TSPNMNDNIL LPVRNNDQA Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 49%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG4L
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUN5
Gene Accession No. Q8TC36
Gene Alias 1700021O15Rik; dJ726C3.1; Sad1 and UNC84 domain containing 5; sad1 and UNC84 domain-containing protein 5; SPAG4L; SPAG4L-2; Sperm-associated antigen 4-like protein; SRG4; SUN domain-containing protein 5; SUN5; testis and spermatogenesis related gene 4; testis and spermatogenesis-related gene 4 protein; TSARG4
Gene Symbols SUN5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPAG4L. Recombinant protein control fragment (Product #RP-99449).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140732
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.