Learn More
SPAG5 Mouse anti-Human, Clone: OTI3F10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810452
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
A novel gene product, hMAP126, interacts with p29 in the yeast two-hybrid assay. The subcellular distribution of hMAP126 was localized to the mitotic spindle and is phosphorylated by p34(cdc2) kinase. Human MAP126 is likely involved in the functional and dynamic regulation of mitotic spindles.Specifications
| SPAG5 | |
| Monoclonal | |
| Unconjugated | |
| SPAG5 | |
| DEEPEST, MAP126, hMAP126 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10615 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3F10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96R06 | |
| SPAG5 | |
| Human recombit protein fragment corresponding to amino acids 1-297 of human SPAG5 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.