Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPAG7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565719
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565719 Supplier Invitrogen™ Supplier No. PA565719
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDDCRYVMIFK KEFAPSDEEL DSYRRGEEWD PQKAEEKRKL KELAQRQEEE AAQQGPVVVS PASDYKDKYS HLIGKGAAKD AAHMLQANKT YGC Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.

SPAG7 gene ontology annotations related to this gene include nucleic acid binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPAG7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPAG7
Gene Accession No. O75391
Gene Alias 5730443G10; ACRP; FSA-1; Fsa1l; hypothetical protein LOC515086; similar to sperm associated antigen 7; SPAG7; sperm associated antigen 7; sperm-associated antigen 7; Sperm-associated antigen 7 homolog; zgc:66397
Gene Symbols SPAG7
Host Species Rabbit
Immunogen Recombinant Human SPAG7. Recombinant protein control fragment (Product #RP-104943).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9552
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.