Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPAG9 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079793JF646
Description
SPAG9 Polyclonal antibody specifically detects SPAG9 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
SPAG9 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
Cancer/testis antigen 89, c-Jun NH2-terminal kinase-associated leucine zipper protein, C-Jun-amino-terminal kinase-interacting protein 4, CT89JIP4, FLJ14006, FLJ26141, FLJ34602, HLC4, HLC-6, HSS, Human lung cancer oncogene 6 protein, JLPPHETSYD1FLJ13450, JNK interacting protein, JNK/SAPK-associated protein, JNK-associated leucine-zipper protein, JNK-interacting protein 4, KIAA0516JIP-4, lung cancer oncogene 4, MAPK8IP4, Max-binding protein, MGC117291, MGC14967, MGC74461, Mitogen-activated protein kinase 8-interacting protein 4, PIG6, proliferation-inducing gene 6, Proliferation-inducing protein 6, Protein highly expressed in testis, sperm associated antigen 9, Sperm surface protein, Sperm-associated antigen 9, Sperm-specific protein, Sunday driver 1 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human JNK/SAPK-Associated Protien using the numbering given in entry NP_003962.3 (GeneID 9043) | |
0.1 mL | |
Cell Cycle and Replication, Signal Transduction | |
9043 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction