Learn More
Invitrogen™ SPAM1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SPAM1. On mouse sample, this antibody also recognizes HYAL5 and HYAL2.
Specifications
Specifications
| Antigen | SPAM1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SPAM1 |
| Gene Accession No. | P38567, P48794 |
| Gene Alias | AV039194; epididymis secretory sperm binding protein Li 96n; HEL-S-96n; HYA1; HYAL1; HYAL3; HYAL5; hyal-PH20; Hyaluronidase PH-20; hyaluronoglucosaminidase PH-20; MGC26532; PH20; Ph-20; PH-20 hyaluronidase; SPAG15; Spam; SPAM1; Sperm adhesion molecule 1; sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding); sperm surface antigen 2B1; Sperm surface protein PH-20 |
| Gene Symbols | SPAM1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.