Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPAM1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2835790.1ML
Description
SPAM1 Polyclonal specifically detects SPAM1 in Human samples. It is validated for Western Blot.Specifications
SPAM1 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
6677 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
EC 3.2.1.35, HYA1, HYAL1, HYAL3, HYAL5, Hyal-PH20, hyaluronidase PH-20, Hyaluronoglucosaminidase PH-20, MGC26532, PH20, PH-20, SPAG15, Sperm adhesion molecule 1, sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding), Sperm surface protein PH-20 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human SPAM1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction