Learn More
Restek™ Spanner Wrench
SDP
Catalog No. 06611082 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6850/6890/7890 FID Collector Assemblies
Catalog No. For Use With (Equipment)
06-611-082 Agilent 5890/6850/6890/7890 FID Collector Assemblies
1 options
Catalog No. 06-611-082 Supplier Restek™ Supplier No. 22329
Add to Cart
Edge
Add to Cart
For Agilent 5890/6850/6890/7890 FID Collector Assemblies
  • Easily remove the nut from the FID collector without damaging the nut
  • Unique, ergonomic handle is easy to grip
  • Fits all instrument models
Specifications
Type Spanner Wrench
Quantity 1
For Use With (Equipment) Agilent 5890/6850/6890/7890 FID Collector Assemblies