Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPANX Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987665
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7665 Supplier Bioss Supplier No. BS7012R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPANXA1, also named as SPANXA and SPANXA2, is a member of the SPANX family of cancer/testis-associated genes. It encodes differentially expressed testis-specific proteins that localize to various subcellular compartments. On Western blots of human testis proteins, SPANX antibodies reacts with the bands migrating between 15–30 kDa. The SPANX protein may be a useful target in cancer immunotherapy.

Specifications

Antigen SPANX
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPANXA2
Gene Accession No. Q9NS26
Gene Alias Cancer/testis antigen 11.1; cancer/testis antigen family 11, member 1; CT11.1; CT111; NAP-X; Nuclear-associated protein SPAN-Xa; SPANX; SPAN-X; SPANX family member A; SPANX family member A2; SPANX family, member A1; SPANX family, member A2; SPANXA; SPAN-Xa; SPANX-A; SPANXA1; SPANXA2; SPAN-Xb; Sperm protein associated with the nucleus on the X chromosome A; sperm protein associated with the nucleus, X chromosome, family member A1; sperm protein associated with the nucleus, X chromosome, family member A2; sperm protein associated with the nucleus, X-linked, family member A1
Gene Symbols SPANXA1, SPANXA2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SPANXA.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30014, 728712
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.