Learn More
Invitrogen™ SPANXA1 Polyclonal Antibody

Description
SPANXA1 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SPANXA1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | SPANXA1 |
| Gene Accession No. | Q9NS26 |
| Gene Alias | cancer/testis antigen 11.1; cancer/testis antigen family 11, member 1; CT11.1; CT111; NAP-X; Nuclear-associated protein SPAN-Xa; SPANX; SPAN-X; SPANX family member A; SPANX family, member A1; SPANXA; SPAN-Xa; SPANX-A; SPANXA1; SPANXA2; SPAN-Xb; Sperm protein associated with the nucleus on the X chromosome A; sperm protein associated with the nucleus, X chromosome, family member A1; sperm protein associated with the nucleus, X-linked, family member A1 |
| Gene Symbols | SPANXA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.