Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPANXC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP286825
Description
SPANXC Polyclonal specifically detects SPANXC in Human samples. It is validated for Western Blot.Specifications
SPANXC | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
B1, C, cancer/testis antigen 11.3, cancer/testis antigen family 11, member 3, cancer/testis-associated protein CTp11, cancer/testis-associated protein of 11 kD, cancer-testis-associated protein CTp11, CT11.1, CT11.2, CT11.3, CT11.4, CTp11, dJ171K16.1, NAP-X, nuclear-associated protein SPAN-Xc, SPANX, SPANX family, member C, SPAN-Xa, SPANXA, SPANX-A, SPAN-Xb, SPANXB, SPANX-B, SPANX-C, SPAN-Xc protein, SPANX-D, sperm protein associated with the nucleus on the X chromosome C, sperm protein associated with the nucleus, X chromosome, family member C | |
The immunogen is a synthetic peptide directed towards the N-terminal region of SPANXC | |
100 μL | |
Cancer | |
64663 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction