Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPANXC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570461 Supplier Invitrogen™ Supplier No. PA570461
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPANXC
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPANXC
Gene Accession No. Q9NY87
Gene Alias C; cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; cancer-testis-associated protein CTp11; CT11.3; CTp11; nuclear-associated protein SPAN-Xc; SPANX family member C; SPANX family, member C; SPANXC; SPANX-C; SPAN-Xc protein; Sperm protein associated with the nucleus on the X chromosome C; sperm protein associated with the nucleus, X chromosome, family member C
Gene Symbols SPANXC
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human SPANXC.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64663
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.