Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SPANXC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583819
Description
Immunogen sequence: SDPQPAPKKL KTSESSTILV VRYRRN.
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
Specifications
SPANXC | |
Polyclonal | |
Unconjugated | |
SPANXC | |
C; cancer/testis antigen 11.3; cancer/testis antigen family 11, member 3; cancer/testis-associated protein CTp11; cancer/testis-associated protein of 11 kD; cancer-testis-associated protein CTp11; CT11.3; CTp11; nuclear-associated protein SPAN-Xc; SPANX family member C; SPANX family, member C; SPANXC; SPANX-C; SPAN-Xc protein; Sperm protein associated with the nucleus on the X chromosome C; sperm protein associated with the nucleus, X chromosome, family member C | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64663 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NY87 | |
SPANXC | |
Recombinant protein corresponding to Human SPANXC. Recombinant protein control fragment (Product #RP-99048). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction