Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPANXD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584932 Supplier Invitrogen™ Supplier No. PA584932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSPEELVNDH ARKNRINPLQ MEEEE Highest antigen sequence indentity to the following orthologs: Rat - 46%, Mouse - 44%.

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a sperm protein that is associated with the nucleus but, although a role in spermatogenesis is suggested, the specific function of this family member has not yet been determined. Polymorphisms in this gene may be associated with prostate cancer susceptibility. [provided by RefSeq, Apr 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPANXD
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPANXD
Gene Accession No. Q9BXN6
Gene Alias cancer/testis antigen 11.4; cancer/testis antigen family 11, member 4; CT11.4; dJ171K16.1; Nuclear-associated protein SPAN-Xd; nuclear-associated protein SPAN-Xe; SPANX family member D; SPANX family, member D; SPANX family, member E; SPANX-C; SPANXD; SPANX-D; SPANXE; SPANX-E; Sperm protein associated with the nucleus on the X chromosome D; sperm protein associated with the nucleus, X chromosome, family member D; sperm protein associated with the nucleus, X chromosome, family member E
Gene Symbols SPANXD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPANXD. Recombinant protein control fragment (Product #RP-108618).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64648
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.