Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPANXN3 Antibody (OTI4B1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274308IR
Description
SPANXN3 Monoclonal specifically detects SPANXN3 in Human samples. It is validated for Western Blot.Specifications
SPANXN3 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SPANXN3 | |
Full length recombinant protein of human SPANXN3 (NP_001009609) produced in E.coli. | |
0.1 mL | |
139067.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI4B1 | |
Western Blot | |
cancer/testis antigen family 11, member 8, CT11.8, Nuclear-associated protein SPAN-Xn3, SPANX family member N3, SPANX family, member N3, SPANX-N3sperm protein associated with the nucleus on the X chromosome N3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction