Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPANXN3 Antibody (OTI4B1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245921
Description
Ensure accurate, reproducible results in Western Blot
SPANXN3 Monoclonal specifically detects SPANXN3 in Human samples. It is validated for Western Blot.Specifications
SPANXN3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 | |
cancer/testis antigen family 11, member 8, CT11.8, Nuclear-associated protein SPAN-Xn3, SPANX family member N3, SPANX family, member N3, SPANX-N3sperm protein associated with the nucleus on the X chromosome N3 | |
Mouse | |
15.4 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
OTI4B1 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
SPANXN3 | |
Full length human recombinant protein of human SPANXN3 (NP_001009609) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
139067 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction