Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPANXN3 Monoclonal Antibody (1F11)
GREENER_CHOICE

Catalog No. PIMA524522
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524522 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524522 Supplier Invitrogen™ Supplier No. MA524522
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEQPTSSTNG EKTKSPCESN NKKNDEMQEV PNRVLAPEQS LKKTKTSEYP IIFVYYLRKG KKINSNQLEN EQSQENSINP IQKEEDEGVD LSEGSSNEDE DLGPCEGPSK EDKDLDSSEG SSQEDEDLGL SEGSSQDSGE D.

SPANXN3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPANXN3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F11
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene SPANXN3
Gene Accession No. Q5MJ09
Gene Alias cancer/testis antigen family 11, member 8; CT11.8; Nuclear-associated protein SPAN-Xn3; SPANX family member N3; SPANX family, member N3; SPANXN3; SPANX-N3; Sperm protein associated with the nucleus on the X chromosome N3
Gene Symbols SPANXN3
Host Species Mouse
Immunogen SPANXN3 (NP_001009609.1, 1 a.a. ~ 141 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139067
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.