Learn More
Invitrogen™ SPANXN3 Monoclonal Antibody (OTI1A2)

Description
SPANXN3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SPANXN3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1A2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SPANXN3 |
| Gene Accession No. | Q5MJ09 |
| Gene Alias | cancer/testis antigen family 11, member 8; CT11.8; Nuclear-associated protein SPAN-Xn3; SPANX family member N3; SPANX family, member N3; SPANXN3; SPANX-N3; Sperm protein associated with the nucleus on the X chromosome N3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.