Learn More
Invitrogen™ SPANXN4 Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PTSSTNENKM KSPCESNKRK VDKKKKNLHR ASAPEQSLKE TEKAKYPTLV FYCRKNKKRN Highest antigen sequence indentity to the following orthologs: Rat - 31%, Mouse - 28%.
Specifications
Specifications
| Antigen | SPANXN4 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SPANXN4 |
| Gene Accession No. | Q5MJ08 |
| Gene Alias | cancer/testis antigen family 11, member 9; CT11.9; nuclear-associated protein SPAN-Xn4; SPANX family member N4; SPANX family, member N4; SPANXN4; SPANX-N4; Sperm protein associated with the nucleus on the X chromosome N4 |
| Gene Symbols | SPANXN4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.