Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPARC Antibody (001), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290612G
Description
SPARC Monoclonal specifically detects SPARC in Mouse samples. It is validated for Western Blot, ELISA.Specifications
SPARC | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
1 | |
Western Blot, ELISA | |
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SPARC (NP_033268.1; Met1-Ile302). | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
6678 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction