Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPARC Antibody (1), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289712AF700
Description
SPARC Monoclonal antibody specifically detects SPARC in Human samples. It is validated for ELISASpecifications
SPARC | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
1 | |
ELISA | |
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SPARC (NP_003109.1; Met 1-Ile 303). | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
6678 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction