Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPARC Antibody (MM0557-8N38) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
| Antigen | SPARC |
|---|---|
| Clone | MM0557-8N38 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SPARC | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Cycle and Replication | |
| P09486 | |
| 6678 | |
| IgG1 | |
| Protein G purified |
| MM0557-8N38 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| SPARC | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title